
Bromoethane-d5

Synonym(s):
Deuterated bromoethane, Ethyl-d5 bromide, Pentadeuteroethyl bromide
Linear Formula:
CD3CD2Br
CAS Number:
Molecular Weight:
114.00
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing