
Aspartame impurity A

Synonym(s):
(2S-cis)-(−)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid
Empirical Formula (Hill Notation):
C13H14N2O4
CAS Number:
Molecular Weight:
262.26
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing