
2-Ethyl-3-methylpyrazine

Empirical Formula (Hill Notation):
C7H10N2
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing