MilliporeSigma
HPLC Analysis of Asparagine Enantiomers on Astec® CHIROBIOTIC® T (25 °C)
HPLC Analysis of Asparagine Enantiomers on Astec® CHIROBIOTIC® T (25 °C) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
guard cartridge
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Guard Column
Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
standard
Product No.
Description
Pricing
L-Asparagine
BioReagent, suitable for cell culture, suitable for insect cell culture
L-Asparagine
≥98% (HPLC)
D-Asparagine monohydrate
≥99% (TLC)
DL-Asparagine monohydrate
≥99.0% (NT)
D-Asparagine
99%
L-Asparagine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water); [B] methanol; [C] formic acid; (30:70:0.02, A:B:C)
flow rate
1 mL/min
detector
UV, 205 nm
column temp.
25 °C
injection
10 μL
sample
300 μg /mL in 30:70 water:methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC