MilliporeSigma
HPLC Analysis of Aflatoxins on Ascentis® Express C18
HPLC Analysis of Aflatoxins on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 μm particles (53823-U)
mobile phase
[A] 5 mM ammonium formate, pH 2.5 (titrated with formic acid); [B] methanol
gradient
5 to 100% B in 10 min
flow rate
0.4 mL/min
pressure
4714 psi (325 bar)
column temp.
35 °C
detector
APCI (+)
injection
5 μL
sample
0.15 - 30 mg/L in 90:10, water:acetonitrile
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of various toxins.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany