MilliporeSigma
HPLC Analysis of 2-Bromo-3-Methylbutyric Acid Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of 2-Bromo-3-Methylbutyric Acid Enantiomers on Astec® CHIROBIOTIC® R application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® R Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
(S)-(−)-2-Bromo-3-methylbutyric acid
96%
2-Bromo-3-methylbutyric acid
97%
(R)-(+)-2-Bromo-3-methylbutyric acid
96%
CONDITIONS
column
CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 μm particles (13024AST)
mobile phase
[A] 20 mM ammonium acetate; [B] methanol; (15:85, A:B)
(alternate mobile phase of 0.1% ammonium acetate in methanol gives 3.5 and 4.3 min retention)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC