MilliporeSigma
GC/MS/MS Analysis of Pesticides in Chamomile on SLB®-5ms after QuEChERS Cleanup using SupelClean Ultra
GC/MS/MS Analysis of Pesticides in Chamomile on SLB®-5ms after QuEChERS Cleanup using SupelClean™ Ultra suitable for GC/MS
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
accessory
Product No.
Description
Pricing
Certified Vial Kit, Low Adsorption (LA) QsertVial
volume 0.3 mL, QsertVial, amber glass vial, natural PTFE/silicone septa, thread for 9 mm
sample prep
Product No.
Description
Pricing
Supel QuE
Empty Centrifuge Tube with Lid, centrifuge tube volume 50 mL, pk of 50, suitable for EN 15662:2008 per BS
Supel QuE
Non-Buffered Tube 1, pk of 50
Supelclean Ultra 2400
volume 3 mL, pkg of 54
CONDITIONS
sample/matrix
chamomile
extraction tube
Supel Que non-buffered extraction tube (4 mg NaSO4, 1 g NaCl in 12 mL tube) (55294-U)
extraction process
Add 10 mL acetonitrile/IS Extraction Solvent and allow to sit 15 minutes, then shake or vortex 30 sec. Add contents of Supel Que non-buffered extraction tube (55294-U). (Shake by hand immediately to prevent clumping. Shake on shaker for 30 minutes at 2250 rpm, centrifuge for 5 minutes at 4500 rpm. Draw off supernatant for cleanup.)
clean-up tube
Supelclean Ultra 2400, 3 mL (54281-U)
clean-up process
See description text
column
SLB-5ms, 30 m x 0.25 mm I.D. x 0.25 μm (28471-U)
oven
105°C (3 min), 10 °C/min to 130 °C, 4°C/min to 200°C, 8°C/min. to 300°C (10 min)
inj. temp.
programmed: 100 °C (.05 min), to 280 °C at 720°C/min, hold 45 min
detector
MS/MS
MSD interface
300 °C
scan range
MRM
carrier gas
helium, 1.4 mL/min constant flow
injection
2 μL splitless
liner
2 mm ID
sample
QuEChERS extract of chamomile
Description
Analysis Note
QuEChERS is a common method used for extraction of pesticide residues from dry botanicals. The resulting extracts are often very high in background, requiring a rigorous cleanup prior to analysis. The standard SPE cleanup approach uses a dual layer cartridge containing graphitized carbon (GCB) and PSA. In this application, Supelclean Ultra 2400 was used for SPE cleanup of chamomile extracts. Supelclean Ultra offers several advantages over the standard cartridge: (1) smaller size and thus less solvent usage, (2) does not require the use of toluene for elution, and (3) better recovery of pesticides with planar structures.
Other Notes
Clean-up process:
Cartridge: Top 3 mL Supelclean Ultra 2400 SPE cartridge (54281-U) with ~0.10 g anhydrous sodium sulfate
Condition: 3 mL of acetonitrile
Load: 750 μL of QuEChERS extract
Elute: using 8 mL of acetonitrile containing 0.5% formic acid, apply slight vacuum at end if needed.
Collect eluent in glass or plastic centrifuge tubes (glass preferred)
Concentrate: all eluent extracts in water bath at 45-50°C w/3 psi nitrogen purge to ~50-100 μL.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany
Supel is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC