MilliporeSigma
  • Inhibition of NAADP signalling on reperfusion protects the heart by preventing lethal calcium oscillations via two-pore channel 1 and opening of the mitochondrial permeability transition pore.
Inhibition of NAADP signalling on reperfusion protects the heart by preventing lethal calcium oscillations via two-pore channel 1 and opening of the mitochondrial permeability transition pore.
Cardiovascular research (2015-09-24)
Sean M Davidson, Kirsty Foote, Suma Kunuthur, Raj Gosain, Noah Tan, Richard Tyser, Yong Juan Zhao, Richard Graeff, A Ganesan, Michael R Duchen, Sandip Patel, Derek M Yellon
ABSTRACT
In the heart, a period of ischaemia followed by reperfusion evokes powerful cytosolic Ca(2+) oscillations that can cause lethal cell injury. These signals represent attractive cardioprotective targets, but the underlying mechanisms of genesis are ill-defined. Here, we investigated the role of the second messenger nicotinic acid adenine dinucleotide phosphate (NAADP), which is known in several cell types to induce Ca(2+) oscillations that initiate from acidic stores such as lysosomes, likely via two-pore channels (TPCs, TPC1 and 2). An NAADP antagonist called Ned-K was developed by rational design based on a previously existing scaffold. Ned-K suppressed Ca(2+) oscillations and dramatically protected cardiomyocytes from cell death in vitro after ischaemia and reoxygenation, preventing opening of the mitochondrial permeability transition pore. Ned-K profoundly decreased infarct size in mice in vivo. Transgenic mice lacking the endo-lysosomal TPC1 were also protected from injury. NAADP signalling plays a major role in reperfusion-induced cell death and represents a potent pathway for protection against reperfusion injury.
Sigma-Aldrich
Ned-K, ≥98% (HPLC)