MilliporeSigma
  • New organometallic imines of rhenium(i) as potential ligands of GSK-3β: synthesis, characterization and biological studies.
New organometallic imines of rhenium(i) as potential ligands of GSK-3β: synthesis, characterization and biological studies.
Dalton transactions (Cambridge, England : 2003) (2018-01-05)
Michelle Muñoz-Osses, Fernando Godoy, Angélica Fierro, Alejandra Gómez, Nils Metzler-Nolte
ABSTRACT
Substituted amino-piperazine derivatives were synthesized and used as precursors for the preparation of a series of new organometallic Re(i) imine complexes with the general formula [(η
Sigma-Aldrich
5-Bromovaleronitrile, 98%