MilliporeSigma
  • Resveratrol Attenuates Copper-Induced Senescence by Improving Cellular Proteostasis.
Resveratrol Attenuates Copper-Induced Senescence by Improving Cellular Proteostasis.
Oxidative medicine and cellular longevity (2017-03-11)
Liliana Matos, Alexandra Monteiro Gouveia, Henrique Almeida
ABSTRACT
Copper sulfate-induced premature senescence (CuSO
Sigma-Aldrich
Anti-DNP antibody produced in rabbit, whole antiserum