MilliporeSigma
  • Isopsoralen-mediated suppression of bone marrow adiposity and attenuation of the adipogenic commitment of bone marrow-derived mesenchymal stem cells.
Isopsoralen-mediated suppression of bone marrow adiposity and attenuation of the adipogenic commitment of bone marrow-derived mesenchymal stem cells.
International journal of molecular medicine (2017-02-17)
Jian Wang, Sheng-Fa Li, Ting Wang, Chun-Han Sun, Liang Wang, Min-Jun Huang, Jian Chen, Shao-Wei Zheng, Nan Wang, Ying-Jun Zhang, Tian-Yu Chen
ABSTRACT
Osteoporosis (OP) increases the risk of bone fractures and other complications, and is thus a major clinical problem. In this study, we examined the effect of isopsoralen on the differentiation of bone-derived marrow mesenchymal stem cells (BMSCs) into osteoblasts and adipocytes, as well as bone formation under osteoporotic conditions. Primary femoral BMSCs isolated from C57BL/6 mice were used to evaluate the isopsoralen-mediated regulation of the expression of alkaline phosphatase (ALP), osteocalcin (OCN) and runt-related transcription factor 2 (RUNX2) during osteogenesis 2 weeks. We also examined the expression of peroxisome proliferator-activated receptor γ (PPARγ) and CCAAT/enhancer binding protein β (C/EBPβ) under adipogenic conditions for 1 and 2 weeks. In addition, ovariectomized (OVX) mice were used to examine the effects of isopsoralen on bone formation for 2 months. Finally, mammalian target of rapamycin complex 1 (mTORC1) signaling was examined under osteogenic and adipogenic conditions. We found that following treatment with isopsoralen, the expression levels of ALP, OCN and RUNX2 were upregulated, whereas those of PPARγ and C/EBPβ were downregulated. mTORC1 signaling was also inhibited in vitro and in vivo. In the OVX mice that were intragastrically administered isopsoralen, bone parameters (trabecular thickness, bone volume/total volume and trabecular number) in the distal femoral metaphysis were significantly increased and the adipocyte number was decreased. On the whole, our findings demonstrate that isopsoralen promoted BMSC differentiation into osteoblasts and suppressed differentiation into adipocytes.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−TRITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Sheep IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody