MilliporeSigma
  • Peginterferon-alpha-2a (40 kD): A review of its use in chronic hepatitis B.
Peginterferon-alpha-2a (40 kD): A review of its use in chronic hepatitis B.
Drugs (2009-12-01)
Gillian M Keating
ABSTRACT
Peginterferon-alpha-2a (40 kD) [Pegasys] comprises an inert, branched, 40 kD polyethylene glycol (PEG) moiety attached to interferon-alpha-2a. Subcutaneous peginterferon-alpha-2a (40 kD) is indicated for the treatment of adults with hepatitis B e antigen (HBeAg)-positive or -negative chronic hepatitis B who have compensated liver disease with evidence of viral replication and hepatic inflammation. Subcutaneous peginterferon-alpha-2a (40 kD) has antiviral and immunomodulatory properties and a convenient once-weekly administration schedule. Forty-eight weeks of therapy with peginterferon-alpha-2a (40 kD) with or without lamivudine was more effective than lamivudine alone in achieving a sustained response in patients with HBeAg-positive or -negative chronic hepatitis B. A long-term follow-up study in patients with HBeAg-positive disease who received peginterferon-alpha-2a (40 kD) monotherapy revealed an HBeAg seroconversion rate of 42%, 1 year after the end of treatment. A long-term follow-up study in patients with HBeAg-negative disease who received peginterferon-alpha-2a (40 kD) with or without lamivudine revealed hepatitis B surface antigen (HBsAg) clearance in 12% of patients and inactive chronic hepatitis B in 17% of patients, 5 years after the end of treatment. Various predictors of response may be useful in terms of identifying patients who may be candidates for shorter or longer peginterferon-alpha-2a (40 kD) treatment durations. For example, quantifying serum HBeAg (in HBeAg-positive disease) and HBsAg levels during therapy may be useful. Adverse events typical of the influenza-like symptoms seen with alpha-interferons occurred more frequently in patients with chronic hepatitis B receiving peginterferon-alpha-2a (40 kD) with or without lamivudine than in those receiving lamivudine alone. In conclusion, peginterferon-alpha-2a (40 kD) is a valuable option for the first-line treatment of HBeAg-negative or -positive chronic hepatitis B.
Sigma-Aldrich
Interferon α 2A human, IFN-α 2A, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture