MilliporeSigma
Differential Metabolism of a Two-Carbon Substrate by Members of the
Frontiers in microbiology (2017-12-13)
Lilian C Baeza, Fabiana R da Mata, Laurine L Pigosso, Maristela Pereira, Gustavo H M F de Souza, Alexandre S G Coelho, Célia M de Almeida Soares
ABSTRACT
The genus
Sigma-Aldrich
Aniline Blue solution, 2.5% in 2% acetic acid