MilliporeSigma
  • The binding of orally dosed hydrophobic active pharmaceutical ingredients to casein micelles in milk.
The binding of orally dosed hydrophobic active pharmaceutical ingredients to casein micelles in milk.
Journal of dairy science (2017-09-18)
M Cheema, A N Hristov, F M Harte
ABSTRACT
Casein proteins (α
Product Description
Supelco
Flunixin, VETRANAL®, analytical standard
Supelco
5-Hydroxyflunixin, VETRANAL®, analytical standard