MilliporeSigma
  • A mass spectrometry and DFT study of pyrithione complexes with transition metals in the gas phase.
A mass spectrometry and DFT study of pyrithione complexes with transition metals in the gas phase.
Journal of mass spectrometry : JMS (2017-07-26)
Matias Butler, Gabriela M Cabrera
ABSTRACT
2-Mercaptopyridine N-oxide (pyrithione, PTOH) along with several transition metal ions forms coordination compounds displaying notable biological activities. Gas-phase complexes formed between pyrithione and manganese (II), cobalt (II), nickel (II), copper (II), and zinc (II) were investigated by infusion in the electrospray source of a quadrupole-time of flight mass spectrometer. Remarkably, positive ion mode spectra displayed the singly charged metal adduct ion [C
Sigma-Aldrich
2-Mercaptopyridine N-oxide, 99%