MilliporeSigma
  • Inhibitory profiles of spices against free and protein-bound heterocyclic amines of roast beef patties as revealed by ultra-performance liquid chromatography-tandem mass spectrometry and principal component analysis.
Inhibitory profiles of spices against free and protein-bound heterocyclic amines of roast beef patties as revealed by ultra-performance liquid chromatography-tandem mass spectrometry and principal component analysis.
Food & function (2017-09-22)
Jing Chen, Zhiyong He, Fang Qin, Jie Chen, Dongsheng Cao, Fengxian Guo, Maomao Zeng
ABSTRACT
The effects of various levels of chili pepper, Sichuan pepper, and black pepper on the amounts of 17 heterocyclic amines (HAs) from seven categories of both free and protein-bound states in roast beef patties were assessed by ultra-performance liquid chromatography-tandem mass spectrometry combined with principal component analysis. Three groups of HA, including imidazopyridines (DMIP), imidazoquinoxalines (MeIQx and 4,8-MeIQx), and β-carbolines (norharman and harman), were detected and quantified in both their free and protein-bound states, whereas PhIP was detected only in its free state, and imidazoquinolines (IQ, IQ[4,5-b], and MeIQ), α-carbolines (AαC and MeAαC), and phenylpyridines (Phe-P-1) were detected only in their protein-bound states. The results demonstrate that the peppers at all three levels had significant inhibitory effects on free PhIP, DMIP, MeIQx, and 4,8-DiMeIQx and could promote free norharman. Harman was significantly suppressed by chili pepper and black pepper, but enhanced by Sichuan pepper. All 11 protein-bound HAs, with the exception of IQ, IQ[4,5-b], and MeIQx with added chili pepper, were significantly reduced by the three peppers. The total amounts of the free and protein-bound states of all 11 HAs (1692.4 ± 78.9 ng g
Sigma-Aldrich
2-Amino-5-phenylpyridine, AldrichCPR