MilliporeSigma
More Than Gliding: Involvement of GldD and GldG in the Virulence of
Frontiers in microbiology (2017-11-23)
David Pérez-Pascual, Tatiana Rochat, Brigitte Kerouault, Esther Gómez, Fabienne Neulat-Ripoll, Celine Henry, Edwige Quillet, Jose A Guijarro, Jean F Bernardet, Eric Duchaud
ABSTRACT
A fascinating characteristic of most members of the genus
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
Orotic acid, ≥98% (titration), anhydrous