MilliporeSigma
  • Magnetic Poly(N-isopropylacrylamide) Nanocomposites: Effect of Preparation Method on Antibacterial Properties.
Magnetic Poly(N-isopropylacrylamide) Nanocomposites: Effect of Preparation Method on Antibacterial Properties.
Nanoscale research letters (2017-10-21)
Nhung H A Nguyen, Mohamed S A Darwish, Ivan Stibor, Pavel Kejzlar, Alena Ševců
ABSTRACT
The most challenging task in the preparation of magnetic poly(N-isopropylacrylamide) (Fe
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%