MilliporeSigma
  • Stunted Growth Caused by Blast Disease in Rice Seedlings Is Associated with Changes in Phytohormone Signaling Pathways.
Stunted Growth Caused by Blast Disease in Rice Seedlings Is Associated with Changes in Phytohormone Signaling Pathways.
Frontiers in plant science (2017-09-22)
Chang-Jie Jiang, Xiao-Long Liu, Xin-Qiong Liu, Hui Zhang, Ying-Jie Yu, Zheng-Wei Liang
ABSTRACT
In response to pathogen attack, plants prioritize defense reactions generally at the expense of plant growth. In this work, we report that changes in phytohormone signaling pathways are associated with the stunted plant growth caused by blast disease in rice seedlings. Infection of rice seedlings with blast fungus
Sigma-Aldrich
Brassinolide, ≥80% (TLC)