MilliporeSigma
  • Hydrogen peroxide in deep waters from the Mediterranean Sea, South Atlantic and South Pacific Oceans.
Hydrogen peroxide in deep waters from the Mediterranean Sea, South Atlantic and South Pacific Oceans.
Scientific reports (2017-03-08)
Mark J Hopwood, Insa Rapp, Christian Schlosser, Eric P Achterberg
ABSTRACT
Hydrogen peroxide (H
Sigma-Aldrich
Vanadium(IV) oxide sulfate hydrate, ≥99.99% trace metals basis