MilliporeSigma
  • Bronchoalveolar lavage fluid from preterm infants with chorioamnionitis inhibits alveolar epithelial repair.
Bronchoalveolar lavage fluid from preterm infants with chorioamnionitis inhibits alveolar epithelial repair.
Respiratory research (2009-11-26)
Jasper V Been, Luc J I Zimmermann, Anne Debeer, Nico Kloosterboer, J Freek van Iwaarden
ABSTRACT
Preterm infants are highly susceptible to lung injury. While both chorioamnionitis and antenatal steroids induce lung maturation, chorioamnionitis is also associated with adverse lung development. We investigated the ability of bronchoalveolar lavage fluid (BALF) from ventilated preterm infants to restore alveolar epithelial integrity after injury in vitro, depending on whether or not they were exposed to chorioamnionitis or antenatal steroids. For this purpose, a translational model for alveolar epithelial repair was developed and characterised. BALF was added to mechanically wounded monolayers of A549 cells. Wound closure was quantified over time and compared between preterm infants (gestational age < 32 wks) exposed or not exposed to chorioamnionitis and antenatal steroids (>or= 1 dose). Furthermore, keratinocyte growth factor (KGF) and vascular endothelial growth factor (VEGF) were quantified in BALF, and their ability to induce alveolar epithelial repair was evaluated in the model. On day 0/1, BALF from infants exposed to antenatal steroids significantly increased epithelial repair (40.3 +/- 35.5 vs. -6.3 +/- 75.0% above control/mg protein), while chorioamnionitis decreased wound-healing capacity of BALF (-2.9 +/- 87.1 vs. 40.2 +/- 36.9% above control/mg protein). BALF from patients with chorioamnionitis contained less KGF (11 (0-27) vs. 0 (0-4) pg/ml) and less detectable VEGF (66 vs. 95%) on day 0. BALF levels of VEGF and KGF correlated with its ability to induce wound repair. Moreover, KGF stimulated epithelial repair dose-dependently, although the low levels in BALF suggest KGF is not a major modulator of BALF-induced wound repair. VEGF also stimulated alveolar epithelial repair, an effect that was blocked by addition of soluble VEGF receptor-1 (sVEGFr1/Flt-1). However, BALF-induced wound repair was not significantly affected by addition of sVEGFr1. Antenatal steroids improve the ability of BALF derived from preterm infants to stimulate alveolar epithelial repair in vitro. Conversely, chorioamnionitis is associated with decreased wound-healing capacity of BALF. A definite role for KGF and VEGF in either process could not be established. Decreased ability to induce alveolar epithelial repair after injury may contribute to the association between chorioamnionitis and adverse lung development in mechanically ventilated preterm infants.
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially fatty acid free