MilliporeSigma
  • Coronary intraplaque hemorrhage evokes a novel atheroprotective macrophage phenotype.
Coronary intraplaque hemorrhage evokes a novel atheroprotective macrophage phenotype.
The American journal of pathology (2009-02-24)
Joseph J Boyle, Heather A Harrington, Emma Piper, Kay Elderfield, Jaroslav Stark, Robert C Landis, Dorian O Haskard
ABSTRACT
Intraplaque hemorrhage accelerates atherosclerosis via oxidant stress and contributes to lesion development and destabilization. Normally, macrophages scavenge hemoglobin-haptoglobin (HbHp) complexes via CD163, and this process provokes the secretion of the anti-inflammatory atheroprotective cytokine interleukin (IL)-10. We therefore tested the hypothesis that HbHp complexes may drive monocyte differentiation to an atheroprotective phenotype. Examination of the macrophage phenotype in hemorrhaged atherosclerotic plaques revealed a novel hemorrhage-associated macrophage population (HA-mac), defined by high levels of CD163, but low levels of human leukocyte antigen-DR. HA-mac contained more iron, a pro-oxidant catalyst, but paradoxically had less oxidative injury, measured by 8-oxo-guanosine content. Differentiating monocytes with HbHp complexes reproduced the CD163(high) human leukocyte antigen-DR(low) HA-mac phenotype in vitro. These in vitro HA-mac cells cleared Hb more quickly, and consistently showed less hydrogen peroxide release, highly reactive oxygen species and oxidant stress, and increased survival. Differentiation to HA-mac was prevented by neutralizing IL-10 antibodies, indicating that IL-10 mediates an autocrine feedback mechanism in this system. Nonlinear dynamic modeling showed that an IL-10/CD163-positive feedback loop drove a discrete HA-mac lineage. Simulations further indicated an all-or-none switch to HA-mac at threshold levels of HbHp, and this conversion was experimentally verified. These data demonstrate the creation of a novel atheroprotective (HA-mac) macrophage subpopulation in response to intraplaque hemorrhage and raise the possibility that therapeutically reproducing this macrophage phenotype may be cardio-protective in cases of atherosclerosis.
Sigma-Aldrich
Anti-8-Hydroxyguanosine Goat pAb, liquid, Calbiochem®
Sigma-Aldrich
Monoclonal Anti-FLAG-Peroxidase antibody produced in rat, 2-4 mg/mL, clone 6F7, purified immunoglobulin
Sigma-Aldrich
Haptoglobin Human, Phenotype 1-1, 98-100%, essentially salt-free, lyophilized powder
Sigma-Aldrich
Hemoglobin A0, Ferrous Stabilized human, lyophilized powder