MilliporeSigma
  • Effects of erythropoietin on cycling performance of well trained cyclists: a double-blind, randomised, placebo-controlled trial.
Effects of erythropoietin on cycling performance of well trained cyclists: a double-blind, randomised, placebo-controlled trial.
The Lancet. Haematology (2017-07-04)
Jules A A C Heuberger, Joris I Rotmans, Pim Gal, Frederik E Stuurman, Juliëtte van 't Westende, Titiaan E Post, Johannes M A Daniels, Matthijs Moerland, Peter L J van Veldhoven, Marieke L de Kam, Herman Ram, Olivier de Hon, Jelle J Posthuma, Jacobus Burggraaf, Adam F Cohen
ABSTRACT
Substances that potentially enhance performance (eg, recombinant human erythropoietin [rHuEPO]) are considered doping and are therefore forbidden in sports; however, the scientific evidence behind doping is frequently weak. We aimed to determine the effects of rHuEPO treatment in well trained cyclists on maximal, submaximal, and race performance and on safety, and to present a model clinical study for doping research on other substances. We did this double-blind, randomised, placebo-controlled trial at the Centre for Human Drug Research in Leiden (Netherlands). We enrolled healthy, well trained but non-professional male cyclists aged 18-50 years and randomly allocated (1:1) them to receive abdominal subcutaneous injections of rHuEPO (epoetin β; mean dose 6000 IU per week) or placebo (0·9% NaCl) for 8 weeks. Randomisation was stratified by age groups (18-34 years and 35-50 years), with a code generated by a statistician who was not masked to the study. The primary outcome was exercise performance, measured as maximal power output (Pmax), maximal oxygen consumption VO Between March 7, 2016, and April 13, 2016, we randomly assigned 48 participants to the rHuEPO group (n=24) or the placebo group (n=24). Mean haemoglobin concentration (9·6 mmol/L vs 9·0 mmol/L [estimated difference 0·6, 95% CI 0·4 to 0·8]) and maximal power output (351·55 W vs 341·23 W [10·32, 3·47 to 17·17]), and VO Although rHuEPO treatment improved a laboratory test of maximal exercise, the more clinically relevant submaximal exercise test performance and road race performance were not affected. This study shows that clinical studies with doping substances can be done adequately and safely and are relevant in determining effects of alleged performance-enhancing drugs. Centre for Human Drug Research, Leiden.
Sigma-Aldrich
Phenyl-β-D-galactopyranoside, ≥98% (TLC)