MilliporeSigma
  • Cellular uptake and anti-tumor activity of gemcitabine conjugated with new amphiphilic cell penetrating peptides.
Cellular uptake and anti-tumor activity of gemcitabine conjugated with new amphiphilic cell penetrating peptides.
EXCLI journal (2017-07-12)
Parvin Zakeri-Milani, Samad Mussa Farkhani, Ali Shirani, Samaneh Mohammadi, Javid Shahbazi Mojarrad, Jafar Akbari, Hadi Valizadeh
ABSTRACT
Gemcitabine (Gem) is used as a single agent or in combination with other anticancer agents to treat many types of solid tumors. However, it has many limitations such as a short plasma half-life, dose-limiting toxicities and drug resistance. Cell-penetrating peptides (CPPs) are short peptides which may deliver a large variety of cargo molecules into the cancerous cells. The current study was designed to evaluate the antiproliferative activity of gemcitabine chemically conjugated to CPPs. The peptides were synthesized using solid phase synthesis procedure. The uptake efficiency of CPPs into cells was examined by flow cytometry and fluorescent microscopy. The synthesized peptides were chemically conjugated to Gem and the
Sigma-Aldrich
O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate, ≥97.0% (N)