MilliporeSigma
A new path to platelet production through matrix sensing.
Haematologica (2017-04-16)
Vittorio Abbonante, Christian Andrea Di Buduo, Cristian Gruppi, Carmelo De Maria, Elise Spedden, Aurora De Acutis, Cristian Staii, Mario Raspanti, Giovanni Vozzi, David L Kaplan, Francesco Moccia, Katya Ravid, Alessandra Balduini
ABSTRACT
Megakaryocytes (MK) in the bone marrow (BM) are immersed in a network of extracellular matrix components that regulates platelet release into the circulation. Combining biological and bioengineering approaches, we found that the activation of transient receptor potential cation channel subfamily V member 4 (TRPV4), a mechano-sensitive ion channel, is induced upon MK adhesion on softer matrices. This response promoted platelet production by triggering a cascade of events that lead to calcium influx, β1 integrin activation and internalization, and Akt phosphorylation, responses not found on stiffer matrices. Lysyl oxidase (LOX) is a physiological modulator of BM matrix stiffness
Sigma-Aldrich
RN-1734, ≥98% (HPLC)
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P5D2, clone P5D2, Chemicon®, from mouse