MilliporeSigma
A TiO
Physical chemistry chemical physics : PCCP (2017-02-06)
Xianfeng Gao, Jianyang Li, Sam Gollon, Ming Qiu, Dongsheng Guan, Xiaoru Guo, Junhong Chen, Chris Yuan
ABSTRACT
The electron transport layer (ETL) plays a critical role in high efficiency perovskite solar cells. In this study, an anodic TiO
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis