MilliporeSigma
  • Large Diversity and Original Structures of Acyl-Homoserine Lactones in Strain MOLA 401, a Marine
Large Diversity and Original Structures of Acyl-Homoserine Lactones in Strain MOLA 401, a Marine
Frontiers in microbiology (2017-07-12)
Margot Doberva, Didier Stien, Jonathan Sorres, Nathalie Hue, Sophie Sanchez-Ferandin, Véronique Eparvier, Yoan Ferandin, Philippe Lebaron, Raphaël Lami
ABSTRACT
Quorum sensing (QS) is a density-dependent mechanism allowing bacteria to synchronize their physiological activities, mediated by a wide range of signaling molecules including