MilliporeSigma
  • Multiphase hydrodechlorination of polychlorinated aromatics - Towards scale-up.
Multiphase hydrodechlorination of polychlorinated aromatics - Towards scale-up.
Chemosphere (2017-02-01)
Alvise Perosa, Maurizio Selva, Thomas Maschmeyer
ABSTRACT
We describe a chemical technology for the reductive catalytic multiphase hydrodechlorination (HDC) of chlorinated aromatics to greatly reduce their toxicity and aid the disposal of such species. The system requires no solvent and the catalyst displays a high recycling efficiency. In the present case, 1,3-dichlorobenzene (1,3-DCB) was used as a model compound, and was quantitatively hydrodechlorinated to benzene with hydrogen, in a tri-phasic liquid system consisting of the chlorinated aromatic itself as the top organic phase, an aqueous sodium hydroxide bottom phase (that neutralises acids formed), and an Aliquat
Sigma-Aldrich
Palladium on activated charcoal, 5% Pd basis
Sigma-Aldrich
Aliquat® 336