MilliporeSigma
  • Formation of Free and Protein-Bound Heterocyclic Amines in Roast Beef Patties Assessed by UPLC-MS/MS.
Formation of Free and Protein-Bound Heterocyclic Amines in Roast Beef Patties Assessed by UPLC-MS/MS.
Journal of agricultural and food chemistry (2017-05-18)
Jing Chen, Zhiyong He, Fang Qin, Jie Chen, Maomao Zeng
ABSTRACT
The effect of different roasting temperatures on the amounts of 17 heterocyclic amines (HAs) from seven categories of both free and protein-bound states in roast beef patties was assessed using an ultraperformance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) method. There were increased amounts and more types of HAs detected at higher roasting temperatures. Nine free HAs were detected at 250 °C, including PhIP (14.34 ± 0.36 ng/g), DMIP (1.02 ± 0.07 ng/g), 1,5,6-TMIP (1.70 ± 0.08 ng/g), MeIQ (0.36 ± 0.01 ng/g), IQx (0.37 ± 0.04 ng/g), MeIQx (9.94 ± 0.61 ng/g), 4,8-DiMeIQx (0.90 ± 0.05 ng/g), norharman (6.03 ± 0.30 ng/g), and harman (2.60 ± 0.09 ng/g). Also, 37.32 ng/g of total free HAs was generated. Twelve protein-bound HAs were detected in roast beef patties at 250 °C, including PhIP (1.70 ± 0.13 ng/g), DMIP (2.33 ± 0.25 ng/g), 1,5,6-TMIP (3.62 ± 0.49 ng/g), MeIQ (5.47 ± 0.18 ng/g), IQ[4,5-b] (0.70 ± 0.03 ng/g) MeIQx (4.03 ± 0.41 ng/g), 4,8-DiMeIQx (0.67 ± 0.09 ng/g), MeAαC (19.51 ± 1.12 ng/g), AαC (2.91 ± 0.45 ng/g), norharman (1304.96 ± 110.73 ng/g), harman (400.85 ± 25.29 ng/g), and Phe-P-1 (0.81 ± 0.06 ng/g). The highest amount of protein-bound HAs was 2913.31 ng/g at 175 °C. PhIP tended to exist in a free state, whereas MeIQ, harman, and norharman tended to exist in a protein-bound state. Furthermore, Phe-P-1, MeAαC, and AαC were detected only in a protein-bound state. These results could be useful for evaluating the exposure to HAs in a daily diet.
Sigma-Aldrich
2-Amino-5-phenylpyridine, AldrichCPR