MilliporeSigma
  • GABAB receptor-mediated regulation of glutamate-activated calcium transients in hypothalamic and cortical neuron development.
GABAB receptor-mediated regulation of glutamate-activated calcium transients in hypothalamic and cortical neuron development.
Journal of neurophysiology (1999-07-13)
K Obrietan, A N van den Pol
ABSTRACT
In the mature nervous system excitatory neurotransmission mediated by glutamate is balanced by the inhibitory actions of GABA. However, during early development, GABA acting at the ligand-gated GABAA Cl- channel also exerts excitatory actions. This raises a question as to whether GABA can exert inhibitory activity during early development, possibly by a mechanism that involves activation of the G protein-coupled GABAB receptor. To address this question we used Ca2+ digital imaging to assess the modulatory role of GABAB receptor signaling in relation to the excitatory effects of glutamate during hypothalamic and cortical neuron development. Ca2+ transients mediated by synaptic glutamate release in neurons cultured from embryonic rat were dramatically depressed by the administration of the GABAB receptor agonist baclofen in a dose-dependent manner. The inhibitory effects of GABAB receptor activation persisted for the duration of baclofen administration (>10 min). Preincubation with the Gi protein inhibitor pertussis toxin resulted in a substantial decrease in the inhibitory actions of baclofen, confirming that a Gi-dependent mechanism mediated the effects of the GABAB receptor. Co-administration of the GABAB receptor antagonist 2-hydroxy-saclofen eliminated the inhibitory action of baclofen. Alone, GABAB antagonist application elicited a marked potentiation of Ca2+ transients mediated by glutamatergic neurotransmission, suggesting that tonic synaptic GABA release exerts an inhibitory tone on glutamate receptor-mediated Ca2+ transients via GABAB receptor activation. In the presence of TTX to block action potential-mediated neurotransmitter release, stimulation with exogenously applied glutamate triggered a robust postsynaptic Ca2+ rise that was dramatically depressed (>70% in cortical neurons, >40% in hypothalamic neurons) by baclofen. Together these data suggest both a pre- and postsynaptic component for the modulatory actions of the GABAB receptor. These results indicate a potentially important role for the GABAB receptor as a modulator of the excitatory actions of glutamate in developing neurons.
Sigma-Aldrich
2-Hydroxysaclofen, ≥98% (TLC), solid