MilliporeSigma
  • Snapshot situation of oxidative degradation of the nervous system, kidney, and adrenal glands biomarkers-neuroprostane and dihomo-isoprostanes-urinary biomarkers from infancy to elderly adults.
Snapshot situation of oxidative degradation of the nervous system, kidney, and adrenal glands biomarkers-neuroprostane and dihomo-isoprostanes-urinary biomarkers from infancy to elderly adults.
Redox biology (2017-01-23)
Libia Alejandra García-Flores, Sonia Medina, Pedro Martínez-Hernández, Camille Oger, Jean-Marie Galano, Thierry Durand, Teresa Casas-Pina, Federico Ferreres, Ángel Gil-Izquierdo
ABSTRACT
We analyzed biomarkers of lipid peroxidation of the nervous system -F
Sigma-Aldrich
2-Hydroxymethyl-1,3-propanediol, 97%