MilliporeSigma
  • Role of calcium ions on the removal of haloacetic acids from swimming pool water by nanofiltration: mechanisms and implications.
Role of calcium ions on the removal of haloacetic acids from swimming pool water by nanofiltration: mechanisms and implications.
Water research (2017-01-08)
Linyan Yang, Jin Zhou, Qianhong She, Man Pun Wan, Rong Wang, Victor W-C Chang, Chuyang Y Tang
ABSTRACT
We investigated the removal of haloacetic acids (HAAs) from swimming pool waters (SPWs) by two nanofiltration membranes NF270 and NF90. The strong matrix effect (particularly by Ca
Sigma-Aldrich
Indophenol Blue, Dye content 60 %