MilliporeSigma
  • Regulators of alternative polyadenylation operate at the transition from mitosis to meiosis.
Regulators of alternative polyadenylation operate at the transition from mitosis to meiosis.
Journal of genetics and genomics = Yi chuan xue bao (2017-02-14)
Lingjuan Shan, Chan Wu, Di Chen, Lei Hou, Xin Li, Lixia Wang, Xiao Chu, Yifeng Hou, Zhaohui Wang
ABSTRACT
In the sexually reproductive organisms, gametes are produced by meiosis following a limited mitotic amplification. However, the intrinsic program switching cells from mitotic to meiotic cycle is unclear. Alternative polyadenylation (APA) is a highly conserved means of gene regulation and is achieved by the RNA 3'-processing machinery to generate diverse 3'UTR profiles. In Drosophila spermatogenesis, we observed distinct profiles of transcriptome-wide 3'UTR between mitotic and meiotic cells. In mutant germ cells stuck in mitosis, 3'UTRs of hundreds of genes were consistently shifted. Remarkably, altering the levels of multiple 3'-processing factors disrupted germline's progression to meiosis, indicative of APA's active role in this transition. An RNA-binding protein (RBP) Tut could directly bind 3'UTRs of 3'-processing factors whose expressions were repressed in the presence of Tut-containing complex. Further, we demonstrated that this RBP complex could execute the repression post-transcriptionally by recruiting CCR4/Twin of deadenylation complex. Thus, we propose that an RBP complex regulates the dynamic APA profile to promote the mitosis-to-meiosis transition.
Sigma-Aldrich
Rabbit IgG−Agarose, saline suspension