MilliporeSigma
  • Efficient, chemoselective synthesis of immunomicelles using single-domain antibodies with a C-terminal thioester.
Efficient, chemoselective synthesis of immunomicelles using single-domain antibodies with a C-terminal thioester.
BMC biotechnology (2009-07-22)
Sanne W A Reulen, Ingrid van Baal, Jos M H Raats, Maarten Merkx
ABSTRACT
Classical bioconjugation strategies for generating antibody-functionalized nanoparticles are non-specific and typically result in heterogeneous compounds that can be compromised in activity. Expression systems based on self-cleavable intein domains allow the generation of recombinant proteins with a C-terminal thioester, providing a unique handle for site-specific conjugation using native chemical ligation (NCL). However, current methods to generate antibody fragments with C-terminal thioesters require cumbersome refolding procedures, effectively preventing application of NCL for antibody-mediated targeting and molecular imaging. Targeting to the periplasm of E. coli allowed efficient production of correctly-folded single-domain antibody (sdAb)-intein fusions proteins. On column purification and 2-mercapthoethanesulfonic acid (MESNA)-induced cleavage yielded single-domain antibodies with a reactive C-terminal MESNA thioester in good yields. These thioester-functionalized single-domain antibodies allowed synthesis of immunomicelles via native chemical ligation in a single step. A novel procedure was developed to obtain soluble, well-folded single-domain antibodies with reactive C-terminal thioesters in good yields. These proteins are promising building blocks for the chemoselective functionalization via NCL of a broad range of nanoparticle scaffolds, including micelles, liposomes and dendrimers.
Sigma-Aldrich
Monoclonal Anti-VSV Glycoprotein antibody produced in mouse, clone P5D4, ascites fluid