MilliporeSigma
  • Enhanced interleukin-12 and CD40 ligand activities but reduced Staphylococcus aureus Cowan 1-induced responses suggest a generalized and progressively impaired type 1 cytokine pattern for human schistosomiasis.
Enhanced interleukin-12 and CD40 ligand activities but reduced Staphylococcus aureus Cowan 1-induced responses suggest a generalized and progressively impaired type 1 cytokine pattern for human schistosomiasis.
Infection and immunity (2002-10-16)
Silvia M L Montenegro, Frederico G C Abath, Ana Lúcia C Domingues, Wlademir G Melo, Clarice N L Morais, Eridan M Coutinho, Siddhartha Mahanty, Thomas A Wynn
ABSTRACT
Whole-blood-cell cultures from schistosomiasis patients were stimulated with a variety of T-cell-dependent and T-cell-independent stimuli to determine whether the defect in type 1 cytokine expression observed following helminth infection is associated with alterations in interleukin-12 (IL-12) or CD40 ligand (CD40L) responsiveness. Cultures from uninfected individuals produced abundant gamma interferon in response to Staphylococcus aureus Cowan 1 (SAC), while patients with intestinal and hepatosplenic disease displayed intermediate and weak responses, respectively. Importantly, the decrease in type 1 cytokine expression was not attributed to defects in IL-12- or CD40L-induced activity. Indeed, schistosomiasis patients displayed heightened responses and even produced more biologically active IL-12 when stimulated with SAC and CD40L than did uninfected controls. Finally, additional studies suggested only a partial role for IL-10, since intestinal patients were the only group that overproduced this downregulatory cytokine. Together, these studies demonstrate that the type 1 deficiency in chronic hepatosplenic schistosomiasis is not related to specific defects in IL-12, IL-10, or CD40L activity, although changes in the functional status of antigen-presenting cells appear to be involved.
Sigma-Aldrich
Protein A from Staphylococcus aureus, Insoluble, Cowan Strain, cell suspension