MilliporeSigma
Colorimetric Fluorescent Sensors for Hemoglobin Based on BODIPY Dyes.
Journal of fluorescence (2016-11-01)
Elif Okutan, Sureyya O Tümay, Serkan Yeşilot
ABSTRACT
Colorimetric fluorescent chemosensors 4 and 5 based on mono- and di- styryl borondipyrromethenes (BODIPY) linked methyl malonyl were designed for detection of hemoglobin (HgB). Their sensing behavior toward various analytes (Br
Sigma-Aldrich
2,4-Dimethylpyrrole, 97%
Sigma-Aldrich
Methyl malonyl chloride, purum, ≥97.0% (AT)