MilliporeSigma
  • Online Pyrophosphate Assay for Analyzing Adenylation Domains of Nonribosomal Peptide Synthetases.
Online Pyrophosphate Assay for Analyzing Adenylation Domains of Nonribosomal Peptide Synthetases.
Chembiochem : a European journal of chemical biology (2016-01-12)
Tiia Kittilä, Melanie Schoppet, Max J Cryle
ABSTRACT
Nonribosomal peptide synthetases (NRPSs) produce many important and structurally complex natural products. Because of their architectures, reprogramming NRPSs has long been attempted to access new bioactive compounds. However, detailed characterization of NRPS catalysis and substrate selectivity by adenylation (A) domains is needed to support such efforts. We present a simple coupled NADH/pyrophosphate (PPi  ) detection assay for analyzing A domain catalysis in vitro. PPi formation is coupled to the consumption of NADH by four enzymatic steps and is detected spectroscopically (λ=340 nm) for simple analysis. We demonstrate the effectiveness of this assay with several adenylation domains, including a stand-alone A domain (DltA, cell wall biosynthesis) and an embedded A domain (Tcp10, teicoplanin biosynthesis). Substrate acceptance of the Tcp10 A domain was explored for the first time, thus demonstrating the applicability of the assay for complex, multi-domain NRPSs.
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type III-S, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein
Sigma-Aldrich
α-Glycerophosphate Dehydrogenase from rabbit muscle, Type I, ammonium sulfate suspension, 100-300 units/mg protein