MilliporeSigma
  • Acute stress enhances adult rat hippocampal neurogenesis and activation of newborn neurons via secreted astrocytic FGF2.
Acute stress enhances adult rat hippocampal neurogenesis and activation of newborn neurons via secreted astrocytic FGF2.
eLife (2013-04-20)
Elizabeth D Kirby, Sandra E Muroy, Wayne G Sun, David Covarrubias, Megan J Leong, Laurel A Barchas, Daniela Kaufer
ABSTRACT
Stress is a potent modulator of the mammalian brain. The highly conserved stress hormone response influences many brain regions, particularly the hippocampus, a region important for memory function. The effect of acute stress on the unique population of adult neural stem/progenitor cells (NPCs) that resides in the adult hippocampus is unclear. We found that acute stress increased hippocampal cell proliferation and astrocytic fibroblast growth factor 2 (FGF2) expression. The effect of acute stress occurred independent of basolateral amygdala neural input and was mimicked by treating isolated NPCs with conditioned media from corticosterone-treated primary astrocytes. Neutralization of FGF2 revealed that astrocyte-secreted FGF2 mediated stress-hormone-induced NPC proliferation. 2 weeks, but not 2 days, after acute stress, rats also showed enhanced fear extinction memory coincident with enhanced activation of newborn neurons. Our findings suggest a beneficial role for brief stress on the hippocampus and improve understanding of the adaptive capacity of the brain. DOI:http://dx.doi.org/10.7554/eLife.00362.001.
Sigma-Aldrich
Adult Rat Hippocampal Neural Stem Cells, Adult Rat Hippocampal Neural Stem Cells are self-renewing cells with the capacity to differentiate into neurons, astrocytes & oligodendrocytes.