MilliporeSigma
  • Facile construction of substituted pyrimido[4,5-d]pyrimidones by transformation of enaminouracil.
Facile construction of substituted pyrimido[4,5-d]pyrimidones by transformation of enaminouracil.
Journal of advanced research (2013-03-01)
Wafaa S Hamama, Mohamed A Ismail, Hanaa A Al-Saman, Hanafi H Zoorob
ABSTRACT
The reaction of 6-amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione (1) as a binucleophile with primary aromatic or heterocyclic amines and formaldehyde or aromatic (heterocyclic) aldehydes in a molar ratio (1:1:2) gave the pyrimido[4,5-d]pyrimidin-2,4-dione ring systems 2-5. Treatment of 1 with diamines and formalin in molar ratio (2:1:4) gave the bis-pyrimido[4,5-d]pyrimidin-2,4-diones 6-8. Furthermore, substituted pyrimido[4,5-d]pyrimidin-2,4-diones with uracil derivative 11 or spiro indole 16 were synthesized. Synthesis of pyrimido[4,5-d]pyrimidin-2,4-diones with different substitution at C-5 and C-7 was achieved to give 13 and 18, respectively.
Sigma-Aldrich
6-Amino-1,3-dimethyluracil, 98%