MilliporeSigma
  • Hepatic crown-like structure: a unique histological feature in non-alcoholic steatohepatitis in mice and humans.
Hepatic crown-like structure: a unique histological feature in non-alcoholic steatohepatitis in mice and humans.
PloS one (2013-12-19)
Michiko Itoh, Hideaki Kato, Takayoshi Suganami, Kuniha Konuma, Yoshio Marumoto, Shuji Terai, Hiroshi Sakugawa, Sayaka Kanai, Miho Hamaguchi, Takahiro Fukaishi, Seiichiro Aoe, Kazunari Akiyoshi, Yoshihiro Komohara, Motohiro Takeya, Isao Sakaida, Yoshihiro Ogawa
ABSTRACT
Although macrophages are thought to be crucial for the pathogenesis of chronic inflammatory diseases, how they are involved in disease progression from simple steatosis to non-alcoholic steatohepatitis (NASH) is poorly understood. Here we report the unique histological structure termed "hepatic crown-like structures (hCLS)" in the mouse model of human NASH; melanocortin-4 receptor deficient mice fed a Western diet. In hCLS, CD11c-positive macrophages aggregate to surround hepatocytes with large lipid droplets, which is similar to those described in obese adipose tissue. Histological analysis revealed that hCLS is closely associated with activated fibroblasts and collagen deposition. When treatment with clodronate liposomes effectively depletes macrophages scattered in the liver, with those in hCLS intact, hepatic expression of inflammatory and fibrogenic genes is unaffected, suggesting that hCLS is an important source of inflammation and fibrosis during the progression of NASH. Notably, the number of hCLS is positively correlated with the extent of liver fibrosis. We also observed increased number of hCLS in the liver of non-alcoholic fatty liver disease/NASH patients. Collectively, our data provide evidence that hCLS is involved in the development of hepatic inflammation and fibrosis, thereby suggesting its pathophysiologic role in disease progression from simple steatosis to NASH.
Sigma-Aldrich
Dichloromethylenediphosphonic acid disodium salt