MilliporeSigma
  • Molecular mechanism of cytotoxicity induced by Hsp90-targeted Antp-TPR hybrid peptide in glioblastoma cells.
Molecular mechanism of cytotoxicity induced by Hsp90-targeted Antp-TPR hybrid peptide in glioblastoma cells.
Molecular cancer (2012-08-24)
Tomohisa Horibe, Aya Torisawa, Masayuki Kohno, Koji Kawakami
ABSTRACT
Heat-shock protein 90 (Hsp90) is vital to cell survival under conditions of stress, and binds client proteins to assist in protein stabilization, translocation of polypeptides across cell membranes, and recovery of proteins from aggregates. Therefore, Hsp90 has emerged as an important target for the treatment of cancer. We previously reported that novel Antp-TPR hybrid peptide, which can inhibit the interaction of Hsp90 with the TPR2A domain of Hop, induces selective cytotoxic activity to discriminate between normal and cancer cells both in vitro and in vivo. In this study, we investigated the functional cancer-cell killing mechanism of Antp-TPR hybrid peptide in glioblastoma (GB) cell lines. It was demonstrated that Antp-TPR peptide induced effective cytotoxic activity in GB cells through the loss of Hsp90 client proteins such as p53, Akt, CDK4, and cRaf. Antp-TPR also did not induce the up-regulation of Hsp70 and Hsp90 proteins, although a small-molecule inhibitor of Hsp90, 17-AAG, induced the up-regulation of these proteins. It was also found that Antp-TPR peptide increased the endoplasmic reticulum unfolded protein response, and the cytotoxic activity of this hybrid peptide to GB cells in the endoplasmic reticulum stress condition. These results show that targeting of Hsp90 by Antp-TPR could be an attractive approach to selective cancer-cell killing because no other Hsp90-targeted compounds show selective cytotoxic activity. Antp-TPR might provide potent and selective therapeutic options for the treatment of cancer.
Sigma-Aldrich
Pifithrin-μ, ≥97% (HPLC), solid