MilliporeSigma
  • Application of headspace solid-phase microextraction followed by gas chromatography-mass spectrometry to determine short-chain alkane monocarboxylic acids in aqueous samples.
Application of headspace solid-phase microextraction followed by gas chromatography-mass spectrometry to determine short-chain alkane monocarboxylic acids in aqueous samples.
Analytical and bioanalytical chemistry (2010-11-26)
Anna Banel, Marta Wasielewska, Bogdan Zygmunt
ABSTRACT
In this study, a procedure was developed to determine short-chain alkane monocarboxylic acids (SCMAs) in aqueous samples using headspace solid-phase microextraction (HS-SPME) followed by gas chromatography (GC) coupled with mass spectrometry (MS). A Stabilwax-DA capillary column (30 m × 0.32-mm inner diameter, 0.50-μm film thickness) was used for GC separation and a 60-μm poly(ethylene glycol) fiber was used to isolate SCMAs from water and introduce them into the gas chromatograph. Parameters of HS-SPME, analyte desorption, and GC-MS analysis were selected and an analytical procedure was proposed. Limits of quantitation were on the order of about 0.2 mg L(-1). As an example of the application of the procedure, SCAMs were determined in municipal wastewater at different steps of treatment.
Product Description
Supelco
Vials, screw top with phenolic open-top cap, pre-assembled, volume 15 mL, clear glass vial, O.D. × H 21 mm × 70 mm, tan PTFE/silicone septum