MilliporeSigma
  • Gene silencing mediated by endogenous microRNAs under heat stress conditions in mammalian cells.
Gene silencing mediated by endogenous microRNAs under heat stress conditions in mammalian cells.
PloS one (2014-07-30)
Masashi Fukuoka, Mariko Yoshida, Akiko Eda, Masaki Takahashi, Hirohiko Hohjoh
ABSTRACT
Heat shock, sudden change in temperature, triggers various responses in cells for protecting the cells from such a severe circumstance. Here we investigated gene silencing mediated by endogenous microRNAs (miRNAs) in mammalian cells exposed to a mild hyperthermia, by means of miRNA activity assay using a luciferase reporter gene as well as miRNA expression analysis using a DNA microarray. Our findings indicated that the gene silencing activities involving miRNAs were enhanced without increasing in their expression levels under heat-stress conditions. Additionally, the gene silencing activity appeared to be independent of the cytoprotective action involving heat shock proteins that are immediately activated in heat-shocked cells and that function as molecular chaperons for restoring heat-denatured proteins to normal proteins. Our current findings suggested the possibility that gene silencing involving endogenous miRNAs might play a subsidiary role in heat-shocked cells for an aggressive inhibition of the expression of heat-denatured proteins.
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-HSP90AB1 (Ab-254) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody