MilliporeSigma
  • Foot-and-mouth disease virus persists in the light zone of germinal centres.
Foot-and-mouth disease virus persists in the light zone of germinal centres.
PloS one (2008-10-23)
Nicholas Juleff, Miriam Windsor, Elizabeth Reid, Julian Seago, Zhidong Zhang, Paul Monaghan, Ivan W Morrison, Bryan Charleston
ABSTRACT
Foot-and-mouth disease virus (FMDV) is one of the most contagious viruses of animals and is recognised as the most important constraint to international trade in animals and animal products. Two fundamental problems remain to be understood before more effective control measures can be put in place. These problems are the FMDV "carrier state" and the short duration of immunity after vaccination which contrasts with prolonged immunity after natural infection. Here we show by laser capture microdissection in combination with quantitative real-time reverse transcription polymerase chain reaction, immunohistochemical analysis and corroborate by in situ hybridization that FMDV locates rapidly to, and is maintained in, the light zone of germinal centres following primary infection of naïve cattle. We propose that maintenance of non-replicating FMDV in these sites represents a source of persisting infectious virus and also contributes to the generation of long-lasting antibody responses against neutralising epitopes of the virus.
Sigma-Aldrich
Anti-Integrin αVβ6 Antibody, clone 10D5, azide free, clone 10D5, Chemicon®, from mouse