MilliporeSigma
  • Synthesis and antiviral activity of a series of HIV-1 protease inhibitors with functionality tethered to the P1 or P1' phenyl substituents: X-ray crystal structure assisted design.
Synthesis and antiviral activity of a series of HIV-1 protease inhibitors with functionality tethered to the P1 or P1' phenyl substituents: X-ray crystal structure assisted design.
Journal of medicinal chemistry (1992-05-15)
W J Thompson, P M Fitzgerald, M K Holloway, E A Emini, P L Darke, B M McKeever, W A Schleif, J C Quintero, J A Zugay, T J Tucker
ABSTRACT
By tethering of a polar hydrophilic group to the P1 or P1' substituent of a Phe-based hydroxyethylene isostere, the antiviral potency of a series of HIV protease inhibitors was improved. The optimum enhancement of anti-HIV activity was observed with the 4-morpholinylethoxy substituent. The substituent effect is consistent with a model derived from inhibitor docked in the crystal structure of the native enzyme. An X-ray crystal structure of the inhibited enzyme determined to 2.25 A verifies the modeling predictions.
Sigma-Aldrich
(1R,2S)-(+)-cis-1-Amino-2-indanol, 99%