MilliporeSigma
  • A structure-activity relationship of non-peptide macrocyclic histone deacetylase inhibitors and their anti-proliferative and anti-inflammatory activities.
A structure-activity relationship of non-peptide macrocyclic histone deacetylase inhibitors and their anti-proliferative and anti-inflammatory activities.
Bioorganic & medicinal chemistry (2015-11-21)
Subhasish Tapadar, Shaghayegh Fathi, Idris Raji, Wilson Omesiete, James R Kornacki, Sandra C Mwakwari, Masanori Miyata, Kazunori Mitsutake, Jian-Dong Li, Milan Mrksich, Adegboyega K Oyelere
ABSTRACT
Inhibition of the enzymatic activity of histone deacetylase (HDAC) is a promising therapeutic strategy for cancer treatment and several distinct small molecule histone deacetylase inhibitors (HDACi) have been reported. We have previously identified a new class of non-peptide macrocyclic HDACi derived from 14- and 15-membered macrolide skeletons. In these HDACi, the macrocyclic ring is linked to the zinc chelating hydroxamate moiety through a para-substituted aryl-triazole cap group. To further delineate the depth of the SAR of this class of HDACi, we have synthesized series of analogous compounds and investigated the influence of various substitution patterns on their HDAC inhibitory, anti-proliferative and anti-inflammatory activities. We identified compounds 25b and 38f with robust anti-proliferative activities and compound 26f (IC50 47.2 nM) with superior anti-inflammatory (IC50 88 nM) activity relative to SAHA.
Sigma-Aldrich
3-Bromobenzaldehyde, 97%
Sigma-Aldrich
2-Ethynylbenzyl alcohol, 95%
Sigma-Aldrich
4-Ethynylbenzyl alcohol, 97%