MilliporeSigma
  • Integrative analysis of copy number alteration and gene expression profiling in ovarian clear cell adenocarcinoma.
Integrative analysis of copy number alteration and gene expression profiling in ovarian clear cell adenocarcinoma.
Cancer genetics (2013-06-04)
Chang Ohk Sung, Chel Hun Choi, Young-Hyeh Ko, Hyunjeong Ju, Yoon-La Choi, Nyunsu Kim, So Young Kang, Sang Yun Ha, Kyusam Choi, Duk-Soo Bae, Jeong-Won Lee, Tae-Joong Kim, Sang Yong Song, Byoung-Gie Kim
ABSTRACT
Ovarian clear cell adenocarcinoma (Ov-CCA) is a distinctive subtype of ovarian epithelial carcinoma. In this study, we performed array comparative genomic hybridization (aCGH) and paired gene expression microarray of 19 fresh-frozen samples and conducted integrative analysis. For the copy number alterations, significantly amplified regions (false discovery rate [FDR] q <0.05) were 1q21.3 and 8q24.3, and significantly deleted regions were 3p21.31, 4q12, 5q13.2, 5q23.2, 5q31.1, 7p22.1, 7q11.23, 8p12, 9p22.1, 11p15.1, 12p13.31, 15q11.2, 15q21.2, 18p11.31, and 22q11.21 using the Genomic Identification of Significant Targets in Cancer (GISTIC) analysis. Integrative analysis revealed 94 genes demonstrating frequent copy number alterations (>25% of samples) that correlated with gene expression (FDR <0.05). These genes were mainly located on 8p11.21, 8p21.2-p21.3, 8q22.1, 8q24.3, 17q23.2-q23.3, 19p13.3, and 19p13.11. Among the regions, 8q24.3 was found to contain the most genes (30 of 94 genes) including PTK2. The 8q24.3 region was indicated as the most significant region, as supported by copy number, GISTIC, and integrative analysis. Pathway analysis using differentially expressed genes on 8q24.3 revealed several major nodes, including PTK2. In conclusion, we identified a set of 94 candidate genes with frequent copy number alterations that correlated with gene expression. Specific chromosomal alterations, such as the 8q24.3 gain containing PTK2, could be a therapeutic target in a subset of Ov-CCAs.