MilliporeSigma
  • MAPN: First-in-Class Reagent for Kinetically Resolved Thiol-to-Thiol Conjugation.
MAPN: First-in-Class Reagent for Kinetically Resolved Thiol-to-Thiol Conjugation.
Bioconjugate chemistry (2015-09-04)
Oleksandr Koniev, Sergii Kolodych, Zoljargal Baatarkhuu, Johann Stojko, Jitka Eberova, Jean-Yves Bonnefoy, Sarah Cianférani, Alain Van Dorsselaer, Alain Wagner
ABSTRACT
Thiols are among the most frequently used functional groups in the field of bioconjugation. While there exists a variety of heterobifunctional reagents that allow for coupling thiols to other functions (e.g., amines, carboxylic acids), there is no specific reagent for creating heteroconjugates using two different thiols. In response to the ever-increasing demand for bioconjugation tools, we have developed p-(maleimide)-phenylpropionitrile (MAPN)-an efficient reagent for kinetically resolved thiol-to-thiol coupling. In a comparative study with its closest commercially available analogue, p-phenylenedimaleimide, MAPN has shown substantial advantages for the preparation of thiol-thiol heteroconjugates. Namely, an antibody-drug conjugate (ADC) with mertansine (DM1), conjugated to the cysteine residues of Trastuzumab, was prepared for the first time.
Sigma-Aldrich
APN-Maleimide