MilliporeSigma
  • Protective Profile Involving CD23/IgE-mediated NO Release is a Hallmark of Cutaneous Leishmaniasis Patients from the Xakriabá Indigenous Community in Minas Gerais, Brazil.
Protective Profile Involving CD23/IgE-mediated NO Release is a Hallmark of Cutaneous Leishmaniasis Patients from the Xakriabá Indigenous Community in Minas Gerais, Brazil.
Scandinavian journal of immunology (2015-03-25)
R Carvalho-Gontijo, V Peruhype-Magalhães, M F Costa-Silva, O A Martins-Filho, P F Quaresma, J de Moura Freire, E de Castro Moreno, A Teixeira-Carvalho, C M Ferreira Gontijo
ABSTRACT
In this study, we described, for the first time, specific aspects of an anti-Leishmania immune response in a Brazilian Xakriabá indigenous community. Induction of an intracellular NO pathway, triggered by the binding of IgE to CD23 receptor in IFN-γ/IL-4 cytokines environment, was evaluated in localized cutaneous leishmaniasis (LCL) carriers and positive Montenegro skin test (MST) individuals without skin lesion (MT(+) SL(-)). Our data demonstrated that the higher frequency of CD23(+) CD14(+) monocytes and the increased serum levels of IgE observed in the LCL group were even higher in LCL carriers with late lesions (LCL≥60). Furthermore, patients with LCL presented increased NO production after Leishmania (Viannia) braziliensis stimulation and this NO profile was independent of the time of the lesion (recent LCL<60 or late LCL≥60). We also showed that the increased frequency of IFN-γ(+) and IL-4(+) CD4(+) T cells is related to the MT(+) SL(-) group. The results of biomarker signature curves demonstrated that in the MT(+) SL(-) group, the index signature was characterized by DAF-2T(+) CD14(+)/IL-4(+) CD8(+)/IFN-γ(+) CD4(+)/IL-4(+) CD4(+). On the other hand, the LCL group presented a higher index of DAF-2T(+) CD14(+)/CD23(+) CD14(+)/IL-4(+) CD8(+), associated with a lower index of IFN-γ(+) CD8(+). Considering the time of lesion, data analysis demonstrated that the main differences observed were highlighted in LCL<60 patients, with a higher index of CD23(+) CD14(+), which was also present in LCL≥60 patients. In conclusion, our data suggest that the protective immune response involving CD23-IgE-mediated NO release is a hallmark of patients with LCL. However, in MT(+) SL(-) individuals, another different leishmanicidal mechanism seems to be involved.