MilliporeSigma
  • Ethyl pyruvate inhibits retinal pathogenic neovascularization by downregulating HMGB1 expression.
Ethyl pyruvate inhibits retinal pathogenic neovascularization by downregulating HMGB1 expression.
Journal of diabetes research (2013-12-29)
Yun Mi Lee, Junghyun Kim, Kyuhyung Jo, So Dam Shin, Chan-Sik Kim, Eun Jin Sohn, Seon Gi Kim, Jin Sook Kim
ABSTRACT
Retinal pathogenic angiogenesis in the eyes is a causative factor in retinopathy of prematurity, diabetic retinopathy, and age-related macular degeneration. This study was designed to examine the pathogenic role of the high-mobility group box-1 (HMGB1) protein and the inhibitory effect of ethyl pyruvate (EP), a well-known antioxidant substance, in retinal pathogenic angiogenesis in mice with oxygen-induced retinopathy (OIR), one of the animal models of proliferative ischemic retinopathy. The OIR mouse model was used for our in vivo studies. The mice were exposed to 75% oxygen from postnatal day 7 (P7) to P11, after which the mice were brought to room air and intraperitoneally injected with EP (50 mg/kg, or 100 mg/kg) for five days. At P17, the mice were perfused with fluorescein isothiocyanate-dextran, and flat-mounted retinas were used to measure nonperfused and neovascular tufts. In OIR mice, an intraperitoneal injection of EP reduced the nonperfused retinal area in the treatment group and significantly reduced the retinal neovascular tufts. In addition, EP inhibited the overexpression of HMGB1 in the retinas of OIR mice. These data suggest that EP could serve as an innovative pharmaceutical agent to prevent retinal neovascularization through inhibiting HMGB1 expression.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 40,000